Learn More
Chemglass Life Sciences Tissue Grinder, 15mL, Tenbroeck, 15mm Dia Pestle, 80mm Chamber Lgth, #0 Rubber Stopper
SDP
Catalog No. 501532400
Encompass_Preferred
Tissue Grinder, 15mL, Tenbroeck, 15mm Dia Pestle, 80mm Chamber Lgth, #0 Rubber Stopper. Are of the same design as the Potter Elvehjem style, but the pestle is hollow allowing the addition of ice water to keep samples chilled during the grinding process.
Catalog No. 50-153-2400 Supplier Chemglass Life Sciences Supplier No. CLS500603
Tissue Grinder, 15mL, Tenbroeck, 15mm Dia Pestle, 80mm Chamber Lgth, #0 Rubber Stopper. Are of the same design as the Potter Elvehjem style, but the pestle is hollow allowing the addition of ice water to keep samples chilled during the grinding process.