Learn More
Chemglass Life Sciences Tissue Grinder, 1mL, Potter-Elvehjem, All Glass, 5.0mm Dia Pestle, 55mm Chember Lgth
SDP
Catalog No. 501532385
Encompass_Preferred
Tissue Grinder, 1mL, Potter-Elvehjem, All Glass, 5.0mm Dia Pestle, 55mm Chember Lgth. The rounded shape on the lower portion of the pestle progressively grinds the sample and the homogenate is produced as the sample is forced through the cylindrical section. Generally used with soft tissue such as brain and liver samples.
Catalog No. 50-153-2385 Supplier Chemglass Life Sciences Supplier No. CLS500402
Tissue Grinder, 1mL, Potter-Elvehjem, All Glass, 5.0mm Dia Pestle, 55mm Chember Lgth. The rounded shape on the lower portion of the pestle progressively grinds the sample and the homogenate is produced as the sample is forced through the cylindrical section. Generally used with soft tissue such as brain and liver samples.