Learn More
Fisherbrand™ Tissue Grinder
SCI_ED
Catalog No. S29230 Shop All Fisherbrand Products
Change view
Click to view available options
Capacity (Metric):
10 mL
Catalog No. Capacity (Metric)
S29230 10 mL
1 options
Catalog No. S29230
Tissue grinder with PTFE plunger head
  • Stainless-steel shaft
  • Precision borosilicate glass bore tube
Specifications
Capacity (Metric) 10 mL