Learn More
Chemglass Life Sciences Tissue Grinder, 30mL, Potter-Elvehjem, All Glass, 19mm Dia Pestle, 105mm Chember Lgth
SDP
Catalog No. 501532390
Encompass_Preferred
Tissue Grinder, 30mL, Potter-Elvehjem, All Glass, 19mm Dia Pestle, 105mm Chember Lgth. The rounded shape on the lower portion of the pestle progressively grinds the sample and the homogenate is produced as the sample is forced through the cylindrical section. Generally used with soft tissue such as brain and liver samples.
Catalog No. 50-153-2390 Supplier Chemglass Life Sciences Supplier No. CLS500407
Tissue Grinder, 30mL, Potter-Elvehjem, All Glass, 19mm Dia Pestle, 105mm Chember Lgth. The rounded shape on the lower portion of the pestle progressively grinds the sample and the homogenate is produced as the sample is forced through the cylindrical section. Generally used with soft tissue such as brain and liver samples.