Learn More
Chemglass Life Sciences Tissue Grinder, 35mL, Free Standing
SDP
Catalog No. 501532426
Encompass_Preferred
Tissue Grinder, 35mL, Free Standing. These tissue grinders are engineered for safety and ease of use. The patented closed system design provides the ability to grind the specimen in a sealed container helping to prevent any risk of contact with the sample. The narrow pestle eliminates the problem of sample overflow common with other tissue grinders. The molded-in abrasive surface does not leave any sediment to obstruct the sample during examination. Tissue grinders come fully assembled and gamma sterilized in an easy- to-open package. An extra cap and patient identification label are provided for storage, transport and traceability.
Catalog No. 50-153-2426 Supplier Chemglass Life Sciences Supplier No. CLS5035
Tissue Grinder, 35mL, Free Standing. These tissue grinders are engineered for safety and ease of use. The patented closed system design provides the ability to grind the specimen in a sealed container helping to prevent any risk of contact with the sample. The narrow pestle eliminates the problem of sample overflow common with other tissue grinders. The molded-in abrasive surface does not leave any sediment to obstruct the sample during examination. Tissue grinders come fully assembled and gamma sterilized in an easy- to-open package. An extra cap and patient identification label are provided for storage, transport and traceability.