Learn More
Chemglass Life Sciences Tissue Grinder, 55mL, Tenbroeck, 25mm Dia Pestle, 125mm Chamber Lgth, #4 Rubber Stopper
SDP
Catalog No. 501532402
Encompass_Preferred
Tissue Grinder, 55mL, Tenbroeck, 25mm Dia Pestle, 125mm Chamber Lgth, #4 Rubber Stopper. Are of the same design as the Potter Elvehjem style, but the pestle is hollow allowing the addition of ice water to keep samples chilled during the grinding process.
Catalog No. 50-153-2402 Supplier Chemglass Life Sciences Supplier No. CLS500605
Tissue Grinder, 55mL, Tenbroeck, 25mm Dia Pestle, 125mm Chamber Lgth, #4 Rubber Stopper. Are of the same design as the Potter Elvehjem style, but the pestle is hollow allowing the addition of ice water to keep samples chilled during the grinding process.