Learn More
Chemglass Life Sciences Tissue Grinder, 55mL, Potter-Elvehjem, 25mm Dia Pestle, 125mm Chamber Lgth
SDP
Catalog No. 501532396
Encompass_Preferred
Tissue Grinder, 55mL, Potter-Elvehjem, 25mm Dia Pestle, 125mm Chamber Lgth. Potter-Elvehjem style tissue grinder with a Stainless Steel /PTFE pestle and an un-ground mortar. The pestle's Stainless Steel rod is for use with an overhead stirrer but the accessory "T" handle can be easily attached to assist with manual grinding. Radial serrations in the lower portion of the pestle facilitate the delivery of homogenate into the cylindrical portion of the mortar.
Catalog No. 50-153-2396 Supplier Chemglass Life Sciences Supplier No. CLS500505
Tissue Grinder, 55mL, Potter-Elvehjem, 25mm Dia Pestle, 125mm Chamber Lgth. Potter-Elvehjem style tissue grinder with a Stainless Steel /PTFE pestle and an un-ground mortar. The pestle's Stainless Steel rod is for use with an overhead stirrer but the accessory "T" handle can be easily attached to assist with manual grinding. Radial serrations in the lower portion of the pestle facilitate the delivery of homogenate into the cylindrical portion of the mortar.