Learn More
Chemglass Life Sciences Tissue Grinder, 5mL, Potter-Elvehjem, 10mm Dia Pestle, 65mm Chamber Lgth
SDP
Catalog No. 501532392
Encompass_Preferred
Tissue Grinder, 5mL, Potter-Elvehjem, 10mm Dia Pestle, 65mm Chamber Lgth. Potter-Elvehjem style tissue grinder with a Stainless Steel /PTFE pestle and an un-ground mortar. The pestle's Stainless Steel rod is for use with an overhead stirrer but the accessory "T" handle can be easily attached to assist with manual grinding. Radial serrations in the lower portion of the pestle facilitate the delivery of homogenate into the cylindrical portion of the mortar.
Catalog No. 50-153-2392 Supplier Chemglass Life Sciences Supplier No. CLS500501
Tissue Grinder, 5mL, Potter-Elvehjem, 10mm Dia Pestle, 65mm Chamber Lgth. Potter-Elvehjem style tissue grinder with a Stainless Steel /PTFE pestle and an un-ground mortar. The pestle's Stainless Steel rod is for use with an overhead stirrer but the accessory "T" handle can be easily attached to assist with manual grinding. Radial serrations in the lower portion of the pestle facilitate the delivery of homogenate into the cylindrical portion of the mortar.