Learn More
Chemglass Life Sciences Tissue Grinder, 5mL, Potter-Elvehjem, All Glass, 10mm Dia Pestle, 65mm Chember Lgth
SDP
Catalog No. 501532387
Encompass_Preferred
Tissue Grinder, 5mL, Potter-Elvehjem, All Glass, 10mm Dia Pestle, 65mm Chember Lgth. The rounded shape on the lower portion of the pestle progressively grinds the sample and the homogenate is produced as the sample is forced through the cylindrical section. Generally used with soft tissue such as brain and liver samples.
Catalog No. 50-153-2387 Supplier Chemglass Life Sciences Supplier No. CLS500404
Tissue Grinder, 5mL, Potter-Elvehjem, All Glass, 10mm Dia Pestle, 65mm Chember Lgth. The rounded shape on the lower portion of the pestle progressively grinds the sample and the homogenate is produced as the sample is forced through the cylindrical section. Generally used with soft tissue such as brain and liver samples.