Learn More
Chemglass Life Sciences Tissue Grinder, 5mL, Tenbroeck, 10mm Dia Pestle, 65mm Chamber Lgth, #000 Rubber Stopper
SDP
Catalog No. 501532398
Encompass_Preferred
Tissue Grinder, 5mL, Tenbroeck, 10mm Dia Pestle, 65mm Chamber Lgth, #000 Rubber Stopper. Are of the same design as the Potter Elvehjem style, but the pestle is hollow allowing the addition of ice water to keep samples chilled during the grinding process.
Catalog No. 50-153-2398 Supplier Chemglass Life Sciences Supplier No. CLS500601
Tissue Grinder, 5mL, Tenbroeck, 10mm Dia Pestle, 65mm Chamber Lgth, #000 Rubber Stopper. Are of the same design as the Potter Elvehjem style, but the pestle is hollow allowing the addition of ice water to keep samples chilled during the grinding process.