Learn More
Electron Microscopy Sciences Tissue Grinder, GPI 13-425 cap 0.5 ml
SDP
Catalog No. 501900364
Encompass_Preferred
This unit features a screw threaded neck for use with Wheaton connection. The 13-425 cap has an open top and PTFE faced silicone rubber liner. The clearance between pestle and mortar is 0.002 – 0.006" (0.05 – 0.15 mm). Made from Wheaton 33 low extractable borosilicate glass
Catalog No. 50-190-0364 Supplier Electron Microscopy Sciences Supplier No. 6480205
This unit features a screw threaded neck for use with Wheaton connection. The 13-425 cap has an open top and PTFE faced silicone rubber liner. The clearance between pestle and mortar is 0.002 – 0.006" (0.05 – 0.15 mm). Made from Wheaton 33 low extractable borosilicate glass