Learn More
Chemglass Life Sciences Tissue Grinder, Dounce, 7mL Working Capacity, 13mm Dia Pestle, 82mm Chamber Lgth
SDP
Catalog No. 501532381
Encompass_Preferred
Tissue Grinder, Dounce, 7mL Working Capacity, 13mm Dia Pestle, 82mm Chamber Lgth. The Dounce tissue grinder is used by moving the pestle up and down and is designed for studies where the nucleus has to remain intact Supplied with two pestles that are used in the same tube, the large clearance pestle is for the initial sample reduction while the small clearance pestle is used to form the final homogenate.
Catalog No. 50-153-2381 Supplier Chemglass Life Sciences Supplier No. CLS500302
Tissue Grinder, Dounce, 7mL Working Capacity, 13mm Dia Pestle, 82mm Chamber Lgth. The Dounce tissue grinder is used by moving the pestle up and down and is designed for studies where the nucleus has to remain intact Supplied with two pestles that are used in the same tube, the large clearance pestle is for the initial sample reduction while the small clearance pestle is used to form the final homogenate.