Learn More
Sklar Instruments Forceps, Iris Tissue Forceps, Sklar 4 in., 1x2 Teeth, Straight, Stainless Steel, EA.
SDP
Catalog No. 501118918 Shop All Sklar Instruments Products
Encompass_Preferred
Forceps, Iris Tissue Forceps, Sklar, 4 in., 1x2 Teeth, Straight, Stainless Steel, EA. Sklar Iris Tissue Forceps are most commonly used for grasping and handling tissue in ophthalmic procedures. The narrow tips, however, allow this instrument to be beneficial for use in other microsurgical procedures as well. Iris Tissue Forceps are available with 1X2 teeth in multiple lengths and curvatures. This product is straight and 4 inches in length. Lifetime warranty against all manufacturer defects.
Catalog No. 50-111-8918 Supplier Sklar Instruments Supplier No. 663340
Forceps, Iris Tissue Forceps, Sklar, 4 in., 1x2 Teeth, Straight, Stainless Steel, EA. Sklar Iris Tissue Forceps are most commonly used for grasping and handling tissue in ophthalmic procedures. The narrow tips, however, allow this instrument to be beneficial for use in other microsurgical procedures as well. Iris Tissue Forceps are available with 1X2 teeth in multiple lengths and curvatures. This product is straight and 4 inches in length. Lifetime warranty against all manufacturer defects.