Learn More
Epredia™ Tissue-Loc™ HistoScreen™ Cassettes, Blue
Catalog No. 22045344 Shop All Epredia Products
Change view
Click to view available options
Color:
Blue
Green
White
Yellow
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-045344 White
22-045348 Blue
22-045347 Green
22-045346 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 22-045344 Supplier Epredia™ Supplier No. C1000WH
This item is not returnable. View return policy
Handle small biopsies and fragile specimens with Epredia™ Tissue-Loc™ HistoScreen™ Cassettes, the ultimate cassettes for specimen security.
  • Saves time, offers the greatest specimen security, and provides ease of use
  • Provides an inner chamber lined with a “see-through" mesh for easy identification of specimens
  • Eliminates the need for messy sponges or lens paper
  • Ensures tissues are safely processed as cassettes will not pop open during processing
Specifications
Pore Style Mesh Pore Insert
Color White
Product Type HistoScreen™ Cassette