Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab™ Tissue Margin Marking Dyes
GSA_VASDP

Catalog No. STLSL662YL2
Change view
Click to view available options
Color:
Black
Blue
Green
Orange
Red
Red, Orange, Yellow, Green, Blue, Violet, Black
Violet
Yellow
Format:
Bottle
Set
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Format
STLSL662YL2 Yellow Bottle
STLSL662RD2 Red Bottle
STLSL662OR2 Orange Bottle
STLSL662GR2 Green Bottle
STLSL662BL2 Blue Bottle
STLSL662VT2 Violet Bottle
STLSL662BK2 Black Bottle
STLSL669 Red, Orange, Yellow, Green, Blue, Violet, Black Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
Catalog No. STLSL662YL2 Supplier StatLab™ Supplier No. SL662YL2
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.

Permanent tissue marking dyes formulated to define margins without bleeding or fading.

  • Durable, bright dyes hold permanently through processing and staining procedures
  • Unique lids may be completely removed or conveniently flipped open to prevent drying
  • Available in 7 dye colors

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Capacity 2 oz.
Color Yellow
For Use With (Application) Defining Tissue Margins
Format Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.