Learn More
Electron Microscopy Sciences Tissue Marking Dye Kit, 5 dyes (5 x 2 oz)
SDP
Catalog No. 501931491
Encompass_Preferred
Fast drying dye is ideal for marking tissue biopsies and pathology specimens and can be used with frozen or fixed tissues. It is a specially formulated binding pigment that evenly coats the tissue without penetrating the tissue surface. The marking dyes do not interfere with diagnostic interpretation due to the fact colors are not similar to standard histopathology dyes. It can be used to permanently mark the margins of excised surgical specimens for exact orientation.
Catalog No. 50-193-1491 Supplier Electron Microscopy Sciences Supplier No. 6302005
Fast drying dye is ideal for marking tissue biopsies and pathology specimens and can be used with frozen or fixed tissues. It is a specially formulated binding pigment that evenly coats the tissue without penetrating the tissue surface. The marking dyes do not interfere with diagnostic interpretation due to the fact colors are not similar to standard histopathology dyes. It can be used to permanently mark the margins of excised surgical specimens for exact orientation.