Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab™ Tissue Marking Dye
SDP

Catalog No. STLSL6628BL
Change view
Click to view available options
Color:
Black
Blue
Green
Orange
Red
Violet
Yellow
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
STLSL6628BL Blue
STLSL6628BK Black
STLSL6628GR Green
STLSL6628OR Orange
STLSL6628RD Red
STLSL6628VT Violet
STLSL6628YL Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
Catalog No. STLSL6628BL Supplier StatLab™ Supplier No. SL6628BL
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.

Permanent tissue marking dyes formulated to define margins without bleeding or fading.

  • Proprietary formulation designed to eliminate evaporation and sludgy dyes, with fewer drips and spills
  • Durable, bright dyes hold permanently through processing and staining procedures
  • Unique lids may either be completely removed or conveniently flipped open to prevent drying

Specifications

Color Blue
Product Type Tissue Marking Dye
Quantity 8 oz. Bottle
Solution Type Mordant Dye
For Use With (Application) Tissue Marking
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.