Learn More
StatLab™ Tissue Marking Dye

Description
- Proprietary formulation designed to eliminate evaporation and sludgy dyes, with fewer drips and spills
- Durable, bright dyes hold permanently through processing and staining procedures
- Unique lids may either be completely removed or conveniently flipped open to prevent drying
Specifications
Specifications
| Color | Violet |
| Product Type | Tissue Marking Dye |
| Quantity | 8 oz. Bottle |
| Solution Type | Mordant Dye |
| For Use With (Application) | Tissue Marking |
| Form | Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.