Learn More
Newcomer Supply IPC Blue Biopsy Marking Dye, 1 oz Dropper Bottle
SDP
Catalog No. NC0336030
Encompass
IPC Blue™ Tissue Marking Dyes are a proprietary formula made with Toluidine Blue biopsy marking dye in a 10% neutral buffered formalin. The dark blue color allows better visualization of biopsy uring embedding and cutting. IPC Blue Tissue Marking Dyes do not interfere with any other staining. The IPC Blue Dye is totally replaced when the slide is stained. (When Eosin is used as the marking dye, the tissue section will remain stained with Eosin). IPC Blue Tissue Marking Dyes do not evaporate as quickly as Eosin because it is formalin based instead of alcohol based.
Catalog No. NC0336030 Supplier Newcomer Supply Supplier No. 6899A
IPC Blue™ Tissue Marking Dyes are a proprietary formula made with Toluidine Blue biopsy marking dye in a 10% neutral buffered formalin. The dark blue color allows better visualization of biopsy uring embedding and cutting. IPC Blue Tissue Marking Dyes do not interfere with any other staining. The IPC Blue Dye is totally replaced when the slide is stained. (When Eosin is used as the marking dye, the tissue section will remain stained with Eosin). IPC Blue Tissue Marking Dyes do not evaporate as quickly as Eosin because it is formalin based instead of alcohol based.