Learn More
Electron Microscopy Sciences Tissue Matrices, 4 mm Diameter, Spherical
SDP
Catalog No. 5029358
Encompass_Preferred
The Tissue Matrix is designed to aid the basic research scientist in the free-hand dissection of a wide variety of different tissues and repeatedly slice fresh tissue at intervals as small as 1 mm. The TM's are constructed from high grade aluminum to allow for rapid temperature equilibration, ease of cleaning or sterilization and withstand the rigors of daily use. Channels at 1 mm intervals. Channel width is 0.3 mm.
Size: 4 mm Diameter
Type: Spherical
Catalog No. 50-293-58 Supplier Electron Microscopy Sciences Supplier No. 69015
The Tissue Matrix is designed to aid the basic research scientist in the free-hand dissection of a wide variety of different tissues and repeatedly slice fresh tissue at intervals as small as 1 mm. The TM's are constructed from high grade aluminum to allow for rapid temperature equilibration, ease of cleaning or sterilization and withstand the rigors of daily use. Channels at 1 mm intervals. Channel width is 0.3 mm.
Size: 4 mm Diameter
Type: Spherical