Learn More
Fisherbrand™ Tissue Path™ IV Biopsy Cassettes
Catalog No. 22035411 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Green
White
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-035411 White
22-282255 Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-035411
This item is not returnable. View return policy
Finely perforated to prevent loss of small biopsy specimens during processing
  • Same two-piece molded hinge design as Tissue Path IV Cassettes
  • Three-sided writing surface assures positive identification
  • Resistant to solvents, decalcifying solutions and compatible with microwave techniques
Order Info
  • Packaged in two dispenser boxes, 250 per box
Specifications
Color White
Material Plastic
Product Type Biopsy Cassette