Learn More
Fisherbrand™ Tissue Path™ IV Biopsy Cassettes
Catalog No. 22282255 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Green
Catalog No. Color
22-282255 Green
1 options
This item is not returnable. View return policy
Catalog No. 22-282255
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View Alternate Product
This item is not returnable. View return policy
Finely perforated to prevent loss of small biopsy specimens during processing
  • Same two-piece molded hinge design as Tissue Path IV Cassettes
  • Three-sided writing surface assures positive identification
  • Resistant to solvents, decalcifying solutions and compatible with microwave techniques
Order Info
  • Packaged in two dispenser boxes, 250 per box
Specifications
Color Green
Material Plastic
Product Type Biopsy Cassette