Learn More
Fisherbrand™ Tissue Path™ IV Biopsy Cassettes
Catalog No. 22313733 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Gray
Green
White
Yellow
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-313-733 Yellow
22-035411 White
22-282255 Green
22-282253 Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 22-313-733
Finely perforated to prevent loss of small biopsy specimens during processing
  • Same two-piece molded hinge design as Tissue Path IV Cassettes
  • Three-sided writing surface assures positive identification
  • Resistant to solvents, decalcifying solutions and compatible with microwave techniques
Order Info
  • Packaged in two dispenser boxes, 250 per box
Specifications
Color Yellow
Material Plastic
Product Type Biopsy Cassette