Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Tissue Path™ Microsette™ Single Compartment Biopsy Cassettes

Catalog No. 15182706A Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Blue
Green
Lilac
Orange
Pink
White
Yellow
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
15-182-706A White
15-182-706B Pink
15-182-706C Green
15-182-706E Blue
15-182-706D Yellow
15-182-706J Orange
15-182-706I Lilac
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 15-182-706A
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Tissue Path™ Microsette™ Single Compartment Biopsy Cassettes are innovative, disposable, and make biopsy pads obsolete.

  • Molded from a special high-density polymer
  • Mesh design features more than 2000, 0.26 mm ''pores'' for maximum fluid exchange and drainage
  • One piece, integral lid eliminates the need for separate lids
  • Lids can be opened or closed as many times as needed and relocked securely each time
  • 45° anterior writing surface suitable for use with certain types of labeling instruments
  • Single Compartment Cassette measures 25 x 30 mm

Specifications

Pore Style Mesh
Writing Face Angle 45°
Closure Type Integral Lid
Color White
No. of Compartments 1
Material High Density Polymer
Product Type Biopsy Cassette
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.