Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Tissue Path™ Microsette™ Six Compartment Biopsy Cassettes

Catalog No. 15182705c Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Aqua
Blue
Gray
Green
Pink
White
Yellow
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
15-182-705C Green
15-182-705L Aqua
15-182-705A White
15-182-705B Pink
15-182-705D Yellow
15-182-705E Blue
15-182-705H Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 15-182-705C
Add to Cart
Add to Cart
This item is not returnable. View return policy

Tissue Path™ Microsette™ Six Compartment Biopsy Cassettes are innovative, disposable and make biopsy pads obsolete.

  • Molded from a special high-density polymer
  • Mesh design features more than 2000, 0.26 mm ''pores'' for maximum fluid exchange and drainage
  • One piece, integral lid eliminates the need for separate lids
  • Lids can be opened or closed as many times as needed and relocked securely each time
  • 45° anterior writing surface suitable for use with certain types of labeling instruments
  • Six-Compartment Cassette holds up to six tissue specimens; each one placed in a separate 0.25 x 0.43 in. (7 x 12 mm) numbered compartment

Specifications

Pore Style Mesh
Writing Face Angle 45°
Color Green
No. of Compartments 6
Material High Density Polymer
Product Type Biopsy Cassette
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.