Learn More
Sigma Aldrich Fine Chemicals Biosciences Tissue plasminogen activat1MG
Catalog No. 502914345
Encompass
Tissue plasminogen activator (tPA) is a protease of the S1 family (trypsin family) and is found in a wide variety of mammalian tissues especially endothelial cells. tPA is secreted as a single chain precursor which is cleaved to a two-chain form by plasmin. Supplied in the single chain form with a molecular weight of ~70nbspkDa. The Tissue Plasminogen Activator (tPA) gene is mapped to human chromosome 8p11.21 and encodes a protease of 69nbspkDa majorly containing serine amino acid residue. It is predominantly expressed in endothelial cells.
Catalog No. 50-291-4345 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. T08311MG
Tissue plasminogen activator (tPA) is a protease of the S1 family (trypsin family) and is found in a wide variety of mammalian tissues especially endothelial cells. tPA is secreted as a single chain precursor which is cleaved to a two-chain form by plasmin. Supplied in the single chain form with a molecular weight of ~70nbspkDa. The Tissue Plasminogen Activator (tPA) gene is mapped to human chromosome 8p11.21 and encodes a protease of 69nbspkDa majorly containing serine amino acid residue. It is predominantly expressed in endothelial cells.