Learn More
Sigma Aldrich Fine Chemicals Biosciences Tissue Preparation Solutio
Catalog No. 502904585
Encompass
The Tissue Preparation SOLUTIONon is intended for use with the Extract-N-Amp(TM) Tissue PCR kits REDExtract-N-Amp(TM) Tissue PCR kits and SYBR(R) Green Extract-N-Amp Tissue PCR kits. This product is intended to supplement the kit reagents where there are many extractions.
Catalog No. 50-290-4585 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. T307330ML
The Tissue Preparation SOLUTIONon is intended for use with the Extract-N-Amp(TM) Tissue PCR kits REDExtract-N-Amp(TM) Tissue PCR kits and SYBR(R) Green Extract-N-Amp Tissue PCR kits. This product is intended to supplement the kit reagents where there are many extractions.