Learn More
Electron Microscopy Sciences Tissue Processing Capsule, White
SDP
Supplier:  Electron Microscopy Sciences 62359W
Encompass_Preferred
These one piece plastic capsules are hinged and perfectly aligned, offering convenience and ease of use. Both the top and the bottom of the capsule have an open mesh offering maximum fluid flow during processing. A frosted marking surface assures permanent identification. These capsules are commonly used with the Embedding Ring System. They are available in two sizes and five colors.
To use simply place the tissue in the capsule, close the cover to lock, and the sample is now ready for processing. 29 (dia)x 6 mm deep are for use with biopsy tissue.
Catalog No. 50-293-66