Learn More
Braintree Scientific Tissue Processing Disks, Large , 4"""" dia., 0.25"""" thick, 10/
SDP
Catalog No. 501956063
Encompass
Blue Wax Tissue Processing Disc Makes Tissue Processing Easy & Quick Process tissue using a clean blue wax surface. Layout, pin, & trim tissue. Use for processing, inspecting, & documenting tissue pathology. Blue Wax is specially formulated to provide the proper consistency to allow pins to stick firmly and be easily removed. **Sold for research purposes only and not for human use**
Catalog No. 50-195-6063 Supplier Braintree Scientific Supplier No. TPDL
Blue Wax Tissue Processing Disc Makes Tissue Processing Easy & Quick Process tissue using a clean blue wax surface. Layout, pin, & trim tissue. Use for processing, inspecting, & documenting tissue pathology. Blue Wax is specially formulated to provide the proper consistency to allow pins to stick firmly and be easily removed. **Sold for research purposes only and not for human use**