Learn More
Braintree Scientific Tissue Processing Disks,Medium ,.75"""" dia.,0.25"""" thick,10/p
SDP
Catalog No. 501956064
Encompass
Blue Wax Tissue Processing Disc Makes Tissue Processing Easy & Quick Process tissue using a clean blue wax surface. Layout, pin, & trim tissue. Use for processing, inspecting, & documenting tissue pathology. Blue Wax is specially formulated to provide the proper consistency to allow pins to stick firmly and be easily removed. **Sold for research purposes only and not for human use**
Catalog No. 50-195-6064 Supplier Braintree Scientific Supplier No. TPDM
Blue Wax Tissue Processing Disc Makes Tissue Processing Easy & Quick Process tissue using a clean blue wax surface. Layout, pin, & trim tissue. Use for processing, inspecting, & documenting tissue pathology. Blue Wax is specially formulated to provide the proper consistency to allow pins to stick firmly and be easily removed. **Sold for research purposes only and not for human use**