Learn More
Electron Microscopy Sciences Embra Tissue Processing Grid 6G150, Copper
SDP
Catalog No. 5029361
Encompass_Preferred
Embra electroformed grids are 3.05 mm in diameter and 16 um thick.
Type: 6G150
Material: Copper
Catalog No. 50-293-61 Supplier Electron Microscopy Sciences Supplier No. ETP150CU
Embra electroformed grids are 3.05 mm in diameter and 16 um thick.
Type: 6G150
Material: Copper