Learn More
Electron Microscopy Sciences Embra Tissue Processing Grid 6G200, Nickel
SDP
Catalog No. 5029364
Encompass_Preferred
Embra electroformed grids are 3.05 mm in diameter and 16 um thick.
Type: 6G200
Material: Nickel
Catalog No. 50-293-64 Supplier Electron Microscopy Sciences Supplier No. ETP200NI
Embra electroformed grids are 3.05 mm in diameter and 16 um thick.
Type: 6G200
Material: Nickel