Learn More
Polysciences, Inc. Tissue Tack Microscope Slides Approximately 72 slides per box Slides provide a positively charged surface which permits instant coupling of negatively charged tissue sections
SDP
Catalog No. NC9901171
Encompass_Preferred
Tissue Tack Microscope Slides Approximately 72 slides per box Slides provide a positively charged surface which permits instant coupling of negatively charged tissue sections
Catalog No. NC9901171 Supplier Polysciences, Inc. Supplier No. 242161
Tissue Tack Microscope Slides Approximately 72 slides per box Slides provide a positively charged surface which permits instant coupling of negatively charged tissue sections