Learn More
BioSpec Products Tissue Tearor II 115v 7 mm
SDP
Catalog No. 502992665
Encompass_Preferred
The new Tissue Tearor II is a rotor/stator type homogenizer designed for the rapid homogenization disruption and emulsification of both plant and animal samples in liquid volumes ranging from 1 0 to 1000 ml The Tissue Tearor II is equipped with a digitally controlled variable-speed electric motor ensuring precise operation for various applications Each Tissue Tearor II comes with a probe of your choice and includes both a storage stand and a operating stand for hands-free use
Catalog No. 50-299-2665 Supplier BioSpec Products Supplier No. 982413007
The new Tissue Tearor II is a rotor/stator type homogenizer designed for the rapid homogenization disruption and emulsification of both plant and animal samples in liquid volumes ranging from 1 0 to 1000 ml The Tissue Tearor II is equipped with a digitally controlled variable-speed electric motor ensuring precise operation for various applications Each Tissue Tearor II comes with a probe of your choice and includes both a storage stand and a operating stand for hands-free use