Learn More
Research Products International Corp Tissue Tearor II Vol 1.0-50ml
SDP
Supplier:  Research Products International Corp 9853722
Encompass
The Tissue Tearor II (TM) is a cutting-edge high-speed rotor/stator homogenizer, expertly engineered to handle a wide range of biological, chemical, and industrial samples. Featuring a motor that is 15% more powerful than its predecessor, this advanced system offers unparalleled efficiency and performance for thorough homogenization, emulsification, and dispersion tasks. Equipped with digital speed control and interchangeable probes, the Tissue Tearor II ensures precise, customizable operation to meet diverse laboratory requirements. Its innovative design includes two specialized stands for hands-free operation and secure storage, enhancing convenience and efficiency in any laboratory setting.
Catalog No. 50-286-5866