Learn More
SAKURA FINETEK USA INC Tissue-Tek Accu-Edge Trimming Blade, Extremely Sharp Edge, Quick Blade Replacement, Disposable
Catalog No. NC2586054
Encompass
The Tissue-Tek Accu-Edge Trimming Blade is engineered for precision in tissue sectioning and trimming during surgical procedures. Designed for efficient and quick blade replacement, it enhances laboratory workflow. This disposable steel blade features an extremely sharp edge, ensuring accurate sample preparation. Its compatibility with specialized handles and grossing boards distinguishes it from competitors, making it an essential tool for anatomists and pathologists.
Catalog No. NC2586054 Supplier SAKURA FINETEK USA INC Supplier No. 4785
The Tissue-Tek Accu-Edge Trimming Blade is engineered for precision in tissue sectioning and trimming during surgical procedures. Designed for efficient and quick blade replacement, it enhances laboratory workflow. This disposable steel blade features an extremely sharp edge, ensuring accurate sample preparation. Its compatibility with specialized handles and grossing boards distinguishes it from competitors, making it an essential tool for anatomists and pathologists.