Learn More
Andwin Scientific Cryomold Intrm 25 x 20 x 5mm 100/pk
SDP
Supplier:  Andwin Scientific 4557
Encompass_Preferred
The Cryomold molds produce uniformly shaped, flat-surface specimen block when used with O.C.T. (optimum cutting temperature) compound, which is sold separately, for frozen sectioning. This can fit into Cryobar freezing wells of the Tissue Tek II microtome/cryostat with the use of an adapter. This product measures 25 mm x 20 mm x 5 mm. It is sold in a pack of 100.
Catalog No. NC9511236