Learn More
Andwin Scientific Tissue-Tek® Cryomold® Molds/Adapters, Intermediate Cryomold, Pack of 100
SDP
Catalog No. NC9542860
Encompass_Preferred
The disposable plastic Cryomold molds produce uniformly shaped, flat-surface specimen block when used with O.C.T. (optimum cutting temperature) compound, which is sold separately, for frozen sectioning. This can fit into Cryobar freezing wells of the Tissue Tek II microtome/cryostat with the use of an adapter. This product measures 15 mm x 15 mm x 5 mm.
Catalog No. NC9542860 Supplier Andwin Scientific Supplier No. 4566
The disposable plastic Cryomold molds produce uniformly shaped, flat-surface specimen block when used with O.C.T. (optimum cutting temperature) compound, which is sold separately, for frozen sectioning. This can fit into Cryobar freezing wells of the Tissue Tek II microtome/cryostat with the use of an adapter. This product measures 15 mm x 15 mm x 5 mm.