Learn More
SAKURA FINETEK USA INC Sakura Finetek Accu-Edge Disposable Microtome Blades, High-Profile, Striation-Free, 50 Blades/Pack
Catalog No. NC3365636
Encompass_Preferred
The Sakura Finetek Tissue-Tek Accu-Edge High-Profile Microtome Blades deliver exceptional precision for cutting soft tissue embedded in paraffin blocks. Designed to minimize common cutting issues like striations and distortion, these blades provide automatic ejection for used blades, ensuring enhanced safety and efficiency in lab workflows. Their compatibility with specialized blade holders and PTFE coating further differentiates them, making them a top choice for histology and pathology professionals.
Catalog No. NC3365636 Supplier SAKURA FINETEK USA INC Supplier No. 468550
The Sakura Finetek Tissue-Tek Accu-Edge High-Profile Microtome Blades deliver exceptional precision for cutting soft tissue embedded in paraffin blocks. Designed to minimize common cutting issues like striations and distortion, these blades provide automatic ejection for used blades, ensuring enhanced safety and efficiency in lab workflows. Their compatibility with specialized blade holders and PTFE coating further differentiates them, making them a top choice for histology and pathology professionals.