Learn More
Andwin Scientific TISSUE-TEK OCT COMPOUND, 4 OZ
SDP
Catalog No. NC9806257
Encompass_Preferred
The Tissue-Tek® optimum cutting temperature (O.C.T.) solution is composed of water-soluble glycols and resins that give a specimen matrix for cryostat sectioning at temperatures of –10°C (14°F) and below. This product does not leave any residue on slides during and after staining prcoess. It is packaged and sold individually in a 118mL (4oz.) bottle.
This item is not returnable. View return policy
Catalog No. NC9806257 Supplier Andwin Scientific Supplier No. 45831EA
The Tissue-Tek® optimum cutting temperature (O.C.T.) solution is composed of water-soluble glycols and resins that give a specimen matrix for cryostat sectioning at temperatures of –10°C (14°F) and below. This product does not leave any residue on slides during and after staining prcoess. It is packaged and sold individually in a 118mL (4oz.) bottle.