Learn More
Andwin Scientific TISSUE TEK STAINING DISH
SDP
Catalog No. NC9670850
Encompass_Preferred
The Tissue-Tek Slide Staining Dish is a replacement for the Tissue-Tek Slide Staining Set, a system specially made for histological or cytological staining procedures. The product is solvent-resistant and is sold in the color of white.
Catalog No. NC9670850 Supplier Andwin Scientific Supplier No. 4457
The Tissue-Tek Slide Staining Dish is a replacement for the Tissue-Tek Slide Staining Set, a system specially made for histological or cytological staining procedures. The product is solvent-resistant and is sold in the color of white.