Learn More
Andwin Scientific Tissue-Tek Slide Staining Dish, White
SDP
Catalog No. NC0731403
Encompass_Preferred
The Tissue-Tek Slide Staining Dish is a replacement for the Tissue-Tek Slide Staining Set, a system specially made for histological or cytological staining procedures. The product is solvent-resistant and is sold in the color of white.
Catalog No. NC0731403 Supplier Andwin Scientific Supplier No. 4457WHITE
The Tissue-Tek Slide Staining Dish is a replacement for the Tissue-Tek Slide Staining Set, a system specially made for histological or cytological staining procedures. The product is solvent-resistant and is sold in the color of white.