Learn More
Andwin Scientific TISSUE-TEK UNI-CASSETTE LILAC
SDP
Catalog No. NC9218341
Encompass_Preferred
The Tissue-Tek Uni-Cassette has a smooth, single action, fracture-back hinge that allows the cassette to fracture when closed, and then break smoothly with no difficulty. This product features a unique design of raised spacer nubs in order to keep the space between the cassettes for better exchange of processing reagents. The cassette also has a wider, thicker finger tab that not only lessens the strain on operators finger, but also makes the opening of the lid less complicated. It also features an oversize writing surface that permits easy organization and identification. This product is sold in the color of lilac.
Catalog No. NC9218341 Supplier Andwin Scientific Supplier No. 62500L
The Tissue-Tek Uni-Cassette has a smooth, single action, fracture-back hinge that allows the cassette to fracture when closed, and then break smoothly with no difficulty. This product features a unique design of raised spacer nubs in order to keep the space between the cassettes for better exchange of processing reagents. The cassette also has a wider, thicker finger tab that not only lessens the strain on operators finger, but also makes the opening of the lid less complicated. It also features an oversize writing surface that permits easy organization and identification. This product is sold in the color of lilac.