Learn More
SAKURA FINETEK USA INC Tissue-Tek Glas Mounting Medium, Xylene-Based Solution, Automated Coverslipping, 500 mL
Catalog No. NC2606967
Encompass
The Tissue-Tek Glas Mounting Medium is a xylene-based solution designed for automated glass coverslipping in histopathology and cytopathology labs. It ensures crystal-clear, bubble-free slides for precise microscopic examination. Engineered for use with the Tissue-Tek Glas g2 Coverslipper, this medium streamlines the coverslipping process and offers high-volume throughput, processing over 16,000 slides per bottle.
Catalog No. NC2606967 Supplier SAKURA FINETEK USA INC Supplier No. 6419
The Tissue-Tek Glas Mounting Medium is a xylene-based solution designed for automated glass coverslipping in histopathology and cytopathology labs. It ensures crystal-clear, bubble-free slides for precise microscopic examination. Engineered for use with the Tissue-Tek Glas g2 Coverslipper, this medium streamlines the coverslipping process and offers high-volume throughput, processing over 16,000 slides per bottle.