Learn More
Covaris Inc tissueTUBE TT1 Plug (25/pack)
SDP
Catalog No. NC9712784
Encompass
Plugs for use with the TT1 and TT1XT. Polypropylene with silicon O-ring for cryogenic storage.
Catalog No. NC9712784 Supplier Covaris Inc Supplier No. 520006
Plugs for use with the TT1 and TT1XT. Polypropylene with silicon O-ring for cryogenic storage.