Learn More
Electron Microscopy Sciences CryoELITE Tissue Vial
SDP
Catalog No. 501929391
Encompass_Preferred
The CryoELITE Tissue Vial is designed specifically for tissue collection, transport, and storage, to provide the utmost protection and insure sample integrity. The tissue vials feature a wide-mouth opening, 5ml capacity and high integrity closure.
Tissue specimens have particular requirements for cryogenic storage. They are manufactured from low binding, cryogenic-grade virgin polypropylene that meets the USP Class VI classification and certified to be free of pyrogens, RNase / DNase and endotoxins, with a storage temperature range of -156 degrees C to 121 degrees C. The externally threaded cap provides a seal that exceeds DOT and IATA classifications for diagnostic specimens and are capable of maintaining a secure closure during freeze/thaw procedures during transport. They have a flat bottom and a stippled external surface to promote easier handling.
Steriel: Sterile
Catalog No. 50-192-9391 Supplier Electron Microscopy Sciences Supplier No. 72616
The CryoELITE Tissue Vial is designed specifically for tissue collection, transport, and storage, to provide the utmost protection and insure sample integrity. The tissue vials feature a wide-mouth opening, 5ml capacity and high integrity closure.
Tissue specimens have particular requirements for cryogenic storage. They are manufactured from low binding, cryogenic-grade virgin polypropylene that meets the USP Class VI classification and certified to be free of pyrogens, RNase / DNase and endotoxins, with a storage temperature range of -156 degrees C to 121 degrees C. The externally threaded cap provides a seal that exceeds DOT and IATA classifications for diagnostic specimens and are capable of maintaining a secure closure during freeze/thaw procedures during transport. They have a flat bottom and a stippled external surface to promote easier handling.
Steriel: Sterile