Learn More
Sigma Aldrich Tissuefab(tm)- Gelma-conductive-vis Bioink 0.2 Um Sterile Filtered Suitable For 3d Bioprinting Applications
Catalog No. 9159631EA Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
91-596-31EA 1 Ea.
1 options
Catalog No. 91-596-31EA Supplier Sigma Aldrich Supplier No. 9159631EA
Only null left
Add to Cart
Add to Cart
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
TissueFab(R)- GelMA-Conductive-Vis bioink is an electrically conductive bioink for 3D bioprinting applications. It is intended to generate ideal cell environments for neural, muscle and cardiac cells, which enable them to transfer electrical signals, enhancing cell communication and capacity for network formation. It is based on gelatin methacryloyl (GelMA) hydrogel system. Carbon nanotubes (CNTs) are incorporated to introduce conductivity, meanwhile enhance the mechanical properties of GelMA hydrogels.TISSUEFAB is a registered trademark of Sigma-Aldrich Co. LLC
Specifications
Quantity 1 Ea.
Synonym 3D Bioprinting; Bioink; GelMA; TissueFab