Learn More
Chemglass Life Sciences Tissue Grinder, 5mL, Griffiths, All Glass, 12mm Dia Pestle, 80mm Chamber Lgth, 150mm OAL
SDP
Catalog No. 501532405
Encompass_Preferred
Tissue Grinder, 5mL, Griffiths, All Glass, 12mm Dia Pestle, 80mm Chamber Lgth, 150mm OAL. The 5mL size is for grinding emulsions and tissues. The ground half round sphere ensure perfect grinding.
Catalog No. 50-153-2405 Supplier Chemglass Life Sciences Supplier No. CLS500801
Tissue Grinder, 5mL, Griffiths, All Glass, 12mm Dia Pestle, 80mm Chamber Lgth, 150mm OAL. The 5mL size is for grinding emulsions and tissues. The ground half round sphere ensure perfect grinding.