Learn More
Chemglass Life Sciences Tissue Grinder, 15mL, Griffiths, All Glass, 15mm Dia Pestle, 95mm Chamber Lgth, 305mm OAL
SDP
Catalog No. 501532406
Encompass_Preferred
Tissue Grinder, 15mL, Griffiths, All Glass, 15mm Dia Pestle, 95mm Chamber Lgth, 305mm OAL. The 15ml size has projections at the end of the pestle for coarse grinding, while the entire length of the chamber is precision ground.
Catalog No. 50-153-2406 Supplier Chemglass Life Sciences Supplier No. CLS500802
Tissue Grinder, 15mL, Griffiths, All Glass, 15mm Dia Pestle, 95mm Chamber Lgth, 305mm OAL. The 15ml size has projections at the end of the pestle for coarse grinding, while the entire length of the chamber is precision ground.