Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab™ TissueGuard™ Biopsy Gel
GSA_VASDP

Catalog No. STLTG12
Change view
Click to view available options
Quantity:
12 x 10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
STLTG12 12 x 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Harmful if swallowed, if in contact with skin, or if inhaled.
Catalog No. STLTG12 Supplier StatLab™ Supplier No. TG12
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Harmful if swallowed, if in contact with skin, or if inhaled.

Stabilize fragile specimens during processing by preparing with TissueGuard™ processing gel.

Cytology: Protect fine needle aspirates, urine specimens, non-gyn specimens, and any other specimens utilizing cell block preparation. Product can be applied in either centrifuge tube, in conjunction with BioMesh tissue cassettes, or using tissue paper. After application, specimen is ready for processing within minutes, or sooner if aided with a cooling block.

Histology: Protect samples including needle biopsies, nerves, small arteries, aggregates, lymph nodes and the smallest of tissue fragments. Product can be applied to samples in conjunction with BioMesh cassettes, or using filter paper.

  • TissueGuard™ encapsulates and retains specimens during histological processing while allowing chemical reagents to penetrate the specimen
  • Appropriate for cytology and histology applications, TissueGuard™ is an alternate to other agarose stabilizers

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Color Amber
Packaging Type Tube
Quantity 12 x 10 mL
For Use With (Application) Stabilizes Specimens For Processing
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.